missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl beta-D-Glucopyranoside 96.0+%, TCI America™

Catalog No. O03551G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O03551G Supplier TCI America Supplier No. O03551G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nonionic surfactant for use in biochemical research.

Chemical Identifiers

CAS 29836-26-8
Molecular Formula C14H28O6
Molecular Weight (g/mol) 292.37
MDL Number MFCD00063288
InChI Key HEGSGKPQLMEBJL-RKQHYHRCSA-N
Synonym octyl-beta-d-glucopyranoside, b-octylglucoside, octyl beta-d-glucopyranoside, octyl beta-d-glucoside, octyl glucoside, n-octyl-beta-d-glucoside, octyl b-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol, unii-v109wut6rl, n-octyl glucoside
PubChem CID 62852
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol
SMILES CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Color White
Quantity 1 g
Formula Weight 292.37
Percent Purity ≥96.0% (GC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material n-Octyl beta-D-Glucopyranoside [for Biochemical Research]
EINECSNumber (5)-6337&(5)-3641
TSCA No