CAS RN 151772-58-6
CAS RN 151772-58-6
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
résultats
1
Chiron Perfluoroalkyl Ether Carboxylic Acid (PFEECAs) Standards
PFAS testing can be highly challenging which is why we make it easier with our convenient portfolio of PFAS reference materials. Advance environmental and health safety with standards you can trust.