CAS RN 144-62-7
Oxalic acid, 98%, anhydrous
CAS: 144-62-7 Molecular Formula: C2H2O4 Molecular Weight (g/mol): 90.034 MDL Number: MFCD00002573 InChI Key: MUBZPKHOEPUJKR-UHFFFAOYSA-N Synonym: ethanedioic acid,aktisal,aquisal,oxiric acid,oxalate,oxalsaeure,oxaalzuur,kyselina stavelova,acide oxalique,acido ossalico PubChem CID: 971 ChEBI: CHEBI:16995 IUPAC Name: oxalic acid SMILES: C(=O)(C(=O)O)O
Oxalic acid, anhydrous, 98%
CAS: 144-62-7 Molecular Formula: C2H2O4 Molecular Weight (g/mol): 90.034 MDL Number: MFCD00002573 InChI Key: MUBZPKHOEPUJKR-UHFFFAOYSA-N Synonym: ethanedioic acid,aktisal,aquisal,oxiric acid,oxalate,oxalsaeure,oxaalzuur,kyselina stavelova,acide oxalique,acido ossalico PubChem CID: 971 ChEBI: CHEBI:16995 IUPAC Name: oxalic acid SMILES: C(=O)(C(=O)O)O
Oxalic acid, 0.1 N standard solution
CAS: 144-62-7 Molecular Formula: C2H2O4 Molecular Weight (g/mol): 90.034 MDL Number: MFCD00002573 InChI Key: MUBZPKHOEPUJKR-UHFFFAOYSA-N Synonym: ethanedioic acid,aktisal,aquisal,oxiric acid,oxalate,oxalsaeure,oxaalzuur,kyselina stavelova,acide oxalique,acido ossalico PubChem CID: 971 ChEBI: CHEBI:16995 IUPAC Name: oxalic acid SMILES: C(=O)(C(=O)O)O
Oxalic acid, 5% w/v aq. soln.
CAS: 144-62-7 Molecular Formula: C2H2O4 Molecular Weight (g/mol): 90.034 MDL Number: MFCD00002573 InChI Key: MUBZPKHOEPUJKR-UHFFFAOYSA-N Synonym: ethanedioic acid,aktisal,aquisal,oxiric acid,oxalate,oxalsaeure,oxaalzuur,kyselina stavelova,acide oxalique,acido ossalico PubChem CID: 971 ChEBI: CHEBI:16995 IUPAC Name: oxalic acid SMILES: C(=O)(C(=O)O)O
Oxalic acid, 0.1N Standardized Solution
CAS: 144-62-7 Molecular Formula: C2H2O4 Molecular Weight (g/mol): 90.034 MDL Number: MFCD00002573 InChI Key: MUBZPKHOEPUJKR-UHFFFAOYSA-N PubChem CID: 971 ChEBI: CHEBI:16995 IUPAC Name: oxalic acid SMILES: C(=O)(C(=O)O)O
Oxalic acid, 10% w/v aq. soln.
CAS: 144-62-7 Molecular Formula: C2H2O4 Molecular Weight (g/mol): 90.034 MDL Number: MFCD00002573 InChI Key: MUBZPKHOEPUJKR-UHFFFAOYSA-N PubChem CID: 971 ChEBI: CHEBI:16995 IUPAC Name: oxalic acid SMILES: C(=O)(C(=O)O)O
Sigma Aldrich Oxalic acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Oxalic Acid Concentrate, 0.005M, Solstice
CAS: 144-62-7 Molecular Formula: C2H2O4 Molecular Weight (g/mol): 90.034 MDL Number: MFCD00002573 InChI Key: MUBZPKHOEPUJKR-UHFFFAOYSA-N Synonym: ethanedioic acid,aktisal,aquisal,oxiric acid,oxalate,oxalsaeure,oxaalzuur,kyselina stavelova,acide oxalique,acido ossalico PubChem CID: 971 ChEBI: CHEBI:16995 IUPAC Name: oxalic acid SMILES: C(=O)(C(=O)O)O
Oxalic Acid Solution, 0.1M, Solstice
CAS: 144-62-7 Molecular Formula: C2H2O4 Molecular Weight (g/mol): 90.034 MDL Number: MFCD00002573 InChI Key: MUBZPKHOEPUJKR-UHFFFAOYSA-N Synonym: ethanedioic acid,aktisal,aquisal,oxiric acid,oxalate,oxalsaeure,oxaalzuur,kyselina stavelova,acide oxalique,acido ossalico PubChem CID: 971 ChEBI: CHEBI:16995 IUPAC Name: oxalic acid SMILES: C(=O)(C(=O)O)O
Oxalic acid solution, Volumetric, 0.5 M (COOH)2 (1N), Solstice
CAS: 144-62-7 Molecular Formula: C2H2O4 Molecular Weight (g/mol): 90.034 InChI Key: MUBZPKHOEPUJKR-UHFFFAOYSA-N Synonym: ethanedioic acid,aktisal,aquisal,oxiric acid,oxalate,oxalsaeure,oxaalzuur,kyselina stavelova,acide oxalique,acido ossalico PubChem CID: 971 ChEBI: CHEBI:16995 IUPAC Name: oxalic acid SMILES: C(=O)(C(=O)O)O
Oxalic Acid Concentrate, 0.05M, Solstice
CAS: 144-62-7 Molecular Formula: C2H2O4 Molecular Weight (g/mol): 90.034 MDL Number: MFCD00002573 InChI Key: MUBZPKHOEPUJKR-UHFFFAOYSA-N Synonym: ethanedioic acid,aktisal,aquisal,oxiric acid,oxalate,oxalsaeure,oxaalzuur,kyselina stavelova,acide oxalique,acido ossalico PubChem CID: 971 ChEBI: CHEBI:16995 IUPAC Name: oxalic acid SMILES: C(=O)(C(=O)O)O
Oxalic Acid Solution, 0.05M, Solstice
CAS: 144-62-7 Molecular Formula: C2H2O4 Molecular Weight (g/mol): 90.034 MDL Number: MFCD00002573 InChI Key: MUBZPKHOEPUJKR-UHFFFAOYSA-N Synonym: ethanedioic acid,aktisal,aquisal,oxiric acid,oxalate,oxalsaeure,oxaalzuur,kyselina stavelova,acide oxalique,acido ossalico PubChem CID: 971 ChEBI: CHEBI:16995 IUPAC Name: oxalic acid SMILES: C(=O)(C(=O)O)O