missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid, 10% w/v aq. soln.

Catalog No. AA35617K2
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35617K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA35617K2 Supplier Thermo Scientific Chemicals Supplier No. 035617K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 144-62-7 | C2H2O4 | 90.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 144-62-7
Molecular Formula C2H2O4
Molecular Weight (g/mol) 90.034
MDL Number MFCD00002573
InChI Key MUBZPKHOEPUJKR-UHFFFAOYSA-N
PubChem CID 971
ChEBI CHEBI:16995
IUPAC Name oxalic acid
SMILES C(=O)(C(=O)O)O

Specifications

Odor Odorless
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Color Colorless
Quantity 1 L
Formula Weight 90.04
Chemical Name or Material Oxalic acid
Hazard Category H290-H318
Precautionary Statement P234-P280-P305+P351+P338-P310-P390
DOTInformation DOT Class: 8; Packing Group: III
EINECSNumber 205-634-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only