missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid, 0.1N Standardized Solution

Catalog No. AA35622K2
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35622K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA35622K2 Supplier Thermo Scientific Chemicals Supplier No. 035622K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 144-62-7 | C2H2O4 | 90.034 g/mol

Oxalic acid, 0.1N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 144-62-7
Molecular Formula C2H2O4
Molecular Weight (g/mol) 90.034
MDL Number MFCD00002573
InChI Key MUBZPKHOEPUJKR-UHFFFAOYSA-N
PubChem CID 971
ChEBI CHEBI:16995
IUPAC Name oxalic acid
SMILES C(=O)(C(=O)O)O

Specifications

Physical Form Liquid
Solubility Miscible with water, alcohol and ether
Quantity 1 L
Formula Weight 90.04
Chemical Name or Material Oxalic acid
EINECSNumber 205-634-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only