missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalic acid, 5% w/v aq. soln.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Concentration | 5% w/v aq. soln. |
| Quantity | 4 L |
| Odor | Odorless |
| Physical Form | Liquid |
| UN Number | UN3265 |
| Formula Weight | 90.04 |
| Chemical Name or Material | Oxalic acid |
Safety and Handling
| Hazard Category | H290-H318 |
| Hazard Statement | GHS H Statement H318-H302-H312 Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. |
| Precautionary Statement | P234-P280-P305+P351+P338-P310-P390 |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S. |
| EINECSNumber | 205-634-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only