missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid concentrate 0.1M (COOH)2 (0.2N), eluent concentrate for IC, MilliporeSigma™ Supelco™

Catalog No. 111013117
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013117 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013117 Supplier Merck Emd Millipore Supplier No. 684871L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 144-62-7
Concentration 0.1 M
Linear Formula HOOCCOOH
MDL Number MFCD00002573
Synonym Oxalic acid solution
Type Organic Standard
Quantity 1 L
Formula Weight 90.03