missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid, 98.0-102.0% (redox titration), MilliporeSigma™ Supelco™

Catalog No. 111013116
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013116 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013116 Supplier Merck Emd Millipore Supplier No. 41706100MG
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 144-62-7
Vapor Pressure <0.01 mmHg (20°C)
Vapor Density 4.4 (vs. air)
Linear Formula HO2 CCO2 H
MDL Number MFCD00002573
Melting Point 189.5°C
Type Analytical Standard
Quantity 100 mg
Formula Weight 90.03
Percent Purity 98.0 to 102.0% (redox titration)
Grade Analytical Standard
Show More Show Less