Learn More
Invitrogen™ Zip Alexa Fluor™ Rapid Antibody Labeling Kits
Invitrogen Zip Alexa Fluor Rapid Antibody Labeling kits allow you to efficiently and quickly label your precious antibody with Alexa Flour dyes to create an antibody conjugate that is ready to use within 15 minutes with 100% recovery.
$638.00 - $638.00
Spécifications
| Kit Contents | Reactive fluorescent label (3 vials), sodium bicarbonate (3 vials) |
|---|---|
| Product Type | Labeling Kit |
| Quantity | 3 Labelings |
| Content And Storage | Store desiccated and protected from light in the freezer (-5°C to +30°C). |
| Labeling Scale | 100 μg |
| Numéro de catalogue | Numéro du manufacturier. | Label or Dye | Excitation/Emission | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Label or Dye | Excitation/Emission | Prix | Quantité | ||||
Z11233
|
Invitrogen™
Z11233 |
Alexa Fluor 488 | 490/525 nm |
chaque for $638.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
Z11234
|
Invitrogen™
Z11234 |
Alexa Fluor 555 | 555/580 nm |
chaque for $638.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Z11235
|
Invitrogen™
Z11235 |
Alexa Fluor 647 | 650/665 nm |
chaque for $638.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
Invitrogen Zip Alexa Fluor Rapid Antibody Labeling kits allow you to efficiently and quickly label your precious antibody with Alexa Flour dyes to create an antibody conjugate that is ready to use within 15 minutes with 100% recovery. Each kit contains everything needed for three separate labeling reactions of 100 μg of antibody. Simply add 1 mL water to component A containing Zip buffer, add 100 μg of antibody, incubate for 15 minutes, and your antibody is ready to use. Conjugates are ideal for multiple applications, including flow cytometry, fluorescent microscopy, immunohistochemistry (IHC), primary detection, ELISAs, immunocytochemistry (ICC), and indirect FISH.
Create ready-to-use antibody conjugate in just 15 minutes with our Invitrogen Zip Alexa Fluor Rapid Antibody Labeling kits. These kits enable you to efficiently label your precious antibody with various Alexa Flour dyes.
Important features of Zip Alexa Fluor Rapid Antibody Labeling kits:
- Speed—antibody conjugates ready to use in 15 min
- Efficiency—100% antibody recovery
- Versatility—kits are useful for a wide range of applications
- Easy-to-use—no chemistry or conjugation experience needed
- Flexibility—choose from Alexa 488, 555, and 647
Each kit contains everything you need to perform three separate labeling reactions of 100 μg of antibody. Conjugates are ideal for multiple applications, including flow cytometry, fluorescent microscopy, immunohistochemistry (IHC), primary detection, ELISA, immunocytochemistry (ICC), and indirect FISH.
Benefits of using Zip Alexa Fluor Rapid Antibody Labeling kits:
Specificity and versatility
A primary antibody that is directly labeled with a fluorophore often produces lower background fluorescence and less nonspecific binding. Multiple primary antibodies of the same isotype or derived from the same species can be used in the same experiment if they are directly labeled with compatible fluorophores.
Robustness
Compared to traditional dyes, Alexa Fluor dyes are brighter, more photostable, and more pH resistant between pH 4 and 10. When using Alexa Fluor dyes, higher degrees of labeling can usually be achieved without intramolecular quenching.
Customization is possible
Can't find what you're looking for in our stocked list? We can prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.
Spécifications
| Reactive fluorescent label (3 vials), sodium bicarbonate (3 vials) | |
| 3 Labelings | |
| 100 μg | |
| Amine | |
| Alexa Fluor |
| Labeling Kit | |
| Store desiccated and protected from light in the freezer (-5°C to +30°C). | |
| Alexa Fluor | |
| IgG |