missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ TSX Series High-Performance Manual Defrost Freezers PROMO
TSX Series high-performance manual defrost freezers offer a sustainable choice for clinical and laboratory storage.
Supplier: Thermo Scientific™ TSX2320FACE
Description
The drive for sample integrity and security- Designed to safely store enzymes, reagents, pharmaceuticals, vaccines, biological and other commonly used laboratory and clinical materials requiring a tight temperature performance
- Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
- Cold wall convection cooling for uniform and stable temperatures
- Easy to use microprocessor control with interior light, setpoint security and alarms
- Standard door locks provide comfort and assurance
- Tested to GMP Clean Room Class A / ISO 5 & 6 (ISO EN 14644-1) with easy to clean surfaces support USP <797> and <800> hazardous drug compounding and other clean room environments
- Tested and certified to conform with the design and safety standards of UL 471, 61010, and CE Directive 2011/65/EU and 2015/863
- Ergonomically designed door handle with a soft-touch grip
- The controller is angled to help prevent neck strain
- Automatic pressure equalization port the enables fast reentry
- Easy to clean and adjust epoxy coated shelves
- Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
- Self-closing door with 90° stop to assist with inventory loads
- US EPA ENERGY STAR Certified with up to 50% less energy usage than conventional refrigerant models*
- ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life.
- Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
- Manufactured in an award-winning, zero-waste to landfill facility
- Thermo Scientific™ DeviceLink™ Connect ready, enabling you to have a connected experience with your unit so you always know the safety of your samples and the performance of your refrigerator or freezer.
- Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems
- Alternate Door swing configuration and door types
- Extra and Alternate wire baskets, shelves, and drawers
- Free-standing or factory-installed chart recorders
- E-Lock Adapter kits
- 4-20 milliamp transmitters and other remote alarm modules
- Specialty, regional, and country specific plugs
- Industry Leading 120-month compressor warranty (varies by country, please contact us for details)
- Optional Certificate of Calibration to an NIST (National Institute of Standards and Technology, formerly National Bureau of Standards) traceable instrument includes a model number and serial number certificate
- Optional Validation Testing Programs include in house and field-based temperature mapping and Installation Qualification (IQ) and Operation Qualification (OQ) programs
- Optional Factory Acceptance Test results including QC Test Log Sheets and QC Temperature Graphs
Spécifications
High-Performance Freezer | |
650 L | |
Solid | |
4 | |
Microprocessor | |
Warranty Varies by Country, Please Contact Us for Details | |
28.5 x 24 x 58 in. (72.3 x 61 x 147.3 cm) | |
37.5 in. | |
95.3 cm | |
78.6 in. | |
199.6 cm | |
28 in. | |
71.1 cm | |
Wireless | |
Smart-Vue Compatible | |
NEMA 5-15 | |
115 V, 60 Hz | |
Energy efficient | |
442 lb. | |
388 lb. | |
8 Racks |
23 cu. ft. | |
-25°C to -15°C (-20°C) | |
1 | |
UL, ENERGYSTAR, cUL | |
TSX Series | |
Manual | |
37.5 x 28 x 78.6 in. (95.3 x 71.1 x 199.6 cm) | |
28.5 in. | |
72.3 cm | |
58 in. | |
147.3 cm | |
24 in. | |
61 cm | |
Optional | |
4.3 A | |
115 V | |
60 Hz | |
R290 | |
200 kg | |
176 kg |
Promotions
-
Thermo Scientific
Rabais jusqu’à 50 % sur ensembles d’entreposage à froid
Jusqu’à -50 % sur équipement d’entreposage à froid Thermo Scientific TSX sélectionné avec ensembles Connect Edge.Expire: 06/30/25
Offre à durée limitée Voir détails promo