missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ ODS-120T HPLC Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™

TSKgel™ ODS-120T columns contain 150 Å pore size silica particles. The exclusion limit of 10,000 Da makes TSKgel™ ODS-120T columns a good choice for the reversed phase chromatography of peptides, small proteins, and small molecular weight compounds in organic and environmental samples.

$3628.34 - $8522.23

Spécifications

Length (Metric) 30 cm
Particle Size 10 μm
pH 2.0 - 7.5
Pore Size 150 Å
Solvents 0 to 100% organic solvent
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Diameter (Metric) Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Diameter (Metric) Prix Quantité  
111021763
Afficher les documents
Merck Emd Millipore
807130
7.8 mm
chaque for $3,628.34
Il en reste null
 
111021764
Afficher les documents
Merck Emd Millipore
807134
21.5 mm
chaque for $8,522.23
Il en reste null
 
Description

Description

The columns are endcapped with trimethyl silane groups to improve the peak shape of positively charged analytes.

Order Info

Shipped in 70% methanol - 30% water.

Spécifications

Spécifications

30 cm
2.0 - 7.5
0 to 100% organic solvent
L1
TSKgel
22%
Stainless Steel
10 μm
150 Å
10°C to 50°C
C18 (octadecyl)
Spherical Silica
Reversed Phase
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'

TSKgel is a registered trademark of Tosoh Corporation