missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TRIS-glycine-SDS running buffer (10X), pH 8.5

TRIS-glycine-SDS running buffer (10X), suitable for protein gel analysis, is used in several research areas, including biochemistry, molecular biology, cell biology, and immunology.

$134.52 - $376.01

Spécifications

Chemical Name or Material TRIS-glycine-SDS running buffer (10X)
Physical Form Liquid
Packaging Plastic bottle
Solubility Information It is soluble in water.
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAJ61006K2
Afficher les documents
Thermo Scientific Chemicals
J61006K2
1 L
chaque for $134.52
Il en reste null
 
AAJ61006K3
Afficher les documents
Thermo Scientific Chemicals
J61006K3
2 L
chaque for $222.42
Il en reste null
 
AAJ61006K7
Afficher les documents
Thermo Scientific Chemicals
J61006K7
4 L
chaque for $376.01
Il en reste null
 
Description

Description

Tris-Glycine SDS Running Buffer (10X) is used as the electrophoresis buffer during the stacking and resolve process of sodium dodecyl sulfate - polyacrylamide gel electrophoresis (SDS-PAGE). It is ideally suited for protein gel analysis in biochemical, molecular biological, cell biological,and immunological research studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• TRIS-glycine-SDS running buffer (10X), is the most commonly used buffer for sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) of proteins

Application

• This solution is the most commonly used buffer for protein gel analysis in several research studies
• It is also used to block membranes with 5% non-fat dry milk for western blotting
• TRIS-glycine-SDS running buffer is commonly used as both anode buffer and cathode buffer
• The use of SDS in the buffer helps keep the proteins linear. Glycine is an amino acid whose charge state plays an important role in the stacking gel

Spécifications

Spécifications

TRIS-glycine-SDS running buffer (10X)
Liquid
Plastic bottle
It is soluble in water.
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only