missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TRIS-buffered saline (TBS, 10X, high salt) pH 7.4

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

$111.93 - $257.74

Identifiants chimiques

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAJ62664AP
Afficher les documents
Thermo Scientific Chemicals
J62664AP
500 mL
chaque for $111.93
Il en reste null
 
AAJ62664K2
Afficher les documents
Thermo Scientific Chemicals
J62664K2
1 L
chaque for $157.11
Il en reste null
 
AAJ62664K3
Afficher les documents
Thermo Scientific Chemicals
J62664K3
2 L
chaque for $257.74
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

77-86-1
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
CHEBI:9754
C(C(CO)(CO)N)O
C4H11NO3
MFCD00004679
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol

Spécifications

77-86-1
7.4
500 mL
MFCD00004679
C(C(CO)(CO)N)O
121.136
CHEBI:9754
10X
Liquid
C4H11NO3
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
TRIS-buffered saline, high salt
Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only