missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TRACE™ TR-1MS GC Columns

Use this low bleed nonpolar column for GC/MS environmental analyses that require high temperature and an inert phase.

$1777.69 - $3330.04

Spécifications

Product Line TRACE
Stationary Phase Trace™ TR-1 MS
USP Type G1, G2, G38
Max. Temperature 340/360°C
Polarity Non-Polar
voir les spécifications

Produits 5
Numéro de catalogue Numéro du manufacturier. Length (Metric) Diameter (Metric) Inner Film Thickness Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Length (Metric) Diameter (Metric) Inner Film Thickness Prix Quantité  
26-0B0-47P
Afficher les documents
Thermo Scientific™
260B047P
30 m 0.25 mm 0.1 μm
chaque for $1,777.69
Il en reste null
 
260B142P
Afficher les documents Détails de la promotion
Thermo Scientific™
260B142P
30 m 0.25 mm 0.25 μm
chaque for $1,985.11
Il en reste null
 
26-0B1-43P
Afficher les documents Détails de la promotion
Thermo Scientific™
260B143P
30 m 0.32 mm 0.25 μm
chaque for $1,852.21
Il en reste null
 
260B154P
Afficher les documents Détails de la promotion
Thermo Scientific™
260B154P
60 m 0.25 mm 0.25 μm
chaque for $3,330.04
Il en reste null
 
26-0B1-55P
Afficher les documents Détails de la promotion
Thermo Scientific™
260B155P
60 m 0.32 mm 0.25 μm
chaque for $3,165.54
Il en reste null
 
Description

Description

With low bleed and a maximum operating temperature of 340–360°C, the Thermo Scientific™ TRACE™ TR-1MS column is ideal for all GC/MS applications. The column is particularly well suited for applications such as GC/MS environmental analyses, which require an inert phase that can operate at high temperature.

Engineered for Superior Performance:
  • Nonpolar phase, 100% dimethyl polysiloxane
  • High operating temperature
  • Inert phase suited for environmental analyses
Applications:
  • Chlorinated and nitroaromatic compounds
  • GC/MS environmental analyses
Excellent Quality and Reproducibility:
  • Low bleed even at elevated temperatures
  • Outstanding robustness for difficult separations
  • Reliable and reproducible performance
Recommended For:
  • Phthalates
  • Polymers
  • Total recoverable petroleum hydrocarbons (TRPHs)
  • Alternative Uses:
  • Chlorinated Aromatics
  • Organochlorine Pesticides
  • Organophosphorous Pesticides
  • PAHs
  • Paraffins
  • Polywax
  • Pyrethroids
  • Semivolatiles
  • Xylenes
Similar To:
  • DB-1
  • DB-Petro
  • BP1
  • HP-1
  • HP-1MS
  • Rtx™-1
  • Ultra-1
  • SPB™-1
  • SPB™-1 Sulfur
  • Petrocol™ DH
  • CP-Sil 5CB
  • RSL-150
  • RSL-160
  • ZB-1
  • CB-1
  • OV™-1
  • PE-1
  • 007-1(MS)
  • SP™-2100
  • SE-30
  • RH-1
  • CC-1
  • CP-Sil 5CB MS
  • VF-1ms
Spécifications

Spécifications

TRACE
G1, G2, G38
Non-Polar
Trace™ TR-1 MS
340/360°C
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'