Learn More
Tians High Performance SMS Lab Coats
Numéro de catalogue. p-3531823
Numéro de catalogue. 191501554 Fournisseur Tians Code fournisseur 864795 M
Il en reste null
Low linting lab coats for use in critical and low particulate clean environments.
  • Constructed of SMS fabric that is strong and durable, yet lightweight and resistant to tears and puncture
  • Features knit collar, left chest pocket and two side pockets with snap front and knit wrist
  • Provides fluid and particulate safety barrier without sacrificing mobility and comfort
  • Impede the spread of viruses, germs, impurities and infections
Spécifications
Product Type Controlled Environments Lab Coat
Material SMS
Color Blue
Closure Type Snaps
Cuff Style Knit
Certifications/Compliance ASTM D751, ASTM D3787, ASTM D2582, ASTM E96-80
Item Description White Wrists, White Collar
Packaging Type Neatly Folded, Cleanroom Packed
Cleanroom Class Class 1000 (ISO 6)