Learn More
Tetraethylammonium perchlorate, 0.2M aqueous solution
CAS: 2567-83-1 | C8H20ClNO4 | 229.70 g/mol
$183.27 - $183.27
Spécifications
Chemical Name or Material | Tetraethylammonium perchlorate |
---|---|
CAS | 2567-83-1,7732-18-5 |
Density | 1.0016g/mL |
Packaging | Plastic bottle |
Molecular Formula | C8H20ClNO4 |
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | |||||
---|---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | |||||
AC379805000
|
Thermo Scientific Chemicals
379805000 |
500 mL | Plastic bottle |
chaque for $183.27
|
|
Vous avez déjà un compte ? Connectez-vous pour voir vos prix et disponibilités. Nouveau ici ? Inscrivez-vous dès aujourd'hui ! |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent, Phase transfer catalystSpécifications
Tetraethylammonium perchlorate | |
1.0016g/mL | |
C8H20ClNO4 | |
1.0016 | |
WGHUNMFFLAMBJD-UHFFFAOYSA-M | |
tetraethylazanium;perchlorate | |
2734932 |
2567-83-1,7732-18-5 | |
Plastic bottle | |
MFCD00041977 | |
tetraethylammonium perchlorate, ethanaminium, n,n,n-triethyl-, perchlorate, tetraethylazanium perchlorate, ethanaminium, n,n,n-triethyl-, perchlorate 1:1, ammonium, tetraethyl-, perchlorate, tetraethylammoniumperchlorate, acmc-1ccq7, ksc203k4p, tetraethyl ammonium perchlorate, tetraethylammoniumperchlorate, dry | |
[O-][Cl](=O)(=O)=O.CC[N+](CC)(CC)CC | |
229.70 | |
229.7 |
RUO – Research Use Only