missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tedizolid

CAS: 856866-72-3 | C17H15FN6O3, C17H15FN6O3 | 370.34 g/mol

160,46 $ - 484,48 $

Identifiants chimiques

CAS 856866-72-3
Molecular Formula C17H15FN6O3
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AC467580010
Afficher les documents
Thermo Scientific Chemicals
467580010
1 g N/A
Il en reste null
 
AC467582500
Afficher les documents
Thermo Scientific Chemicals
467582500
250 mg N/A
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

856866-72-3
C17H15FN6O3

Spécifications

856866-72-3
201.0°C
370.34
CN1N=NC(=N1)C1=NC=C(C=C1)C1=C(F)C=C(C=C1)N1CC(CO)OC1=O
370.34
1 g
C17H15FN6O3
XFALPSLJIHVRKE-UHFFFAOYNA-N
3-{3-fluoro-4-[6-(2-methyl-2H-1,2,3,4-tetrazol-5-yl)pyridin-3-yl]phenyl}-5-(hydroxymethyl)-1,3-oxazolidin-2-one
Tedizolid
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only