missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TA Cloning™ Kit, Dual Promoter, with pCR™II Vector and One Shot™ INVαF' Chemically Competent E. coli DFS Item

Provides quick, one-step cloning strategy for directly inserting a Taq-amplified PCR product into plasmid vector

Supplier:  Invitrogen™ K205040

 Afficher plus de versions de ce produit

Numéro de catalogue. K205040


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

The T7 and Sp6 promoters of the pCRII vector allow in vitro transcription of the insert to produce sense or anti-sense products. The TA Cloning Kit Dual Promoter uses the pCRII cloning vector and ExpressLink T4 DNA Ligase to generate a ligation product in a fifteen-minute, room-temperature ligation step. Reactions typically yield >80% recombinants containing inserts.

  • Fast and convenient: 15-minute, room-temperature ligation
  • Efficient: blue/white screening and >80% clones with correct insert
  • Flexible: choice of kanamycin or ampicillin resistance for flexible antibiotic selection
  • Hassle-free: eliminates any enzymatic modifications of PCR product
  • Streamlined: does not require use of PCR primers that contain restriction sites
  • 3'-T overhangs for direct ligation of Taq-amplified PCR products
  • T7 and Sp6 promoters for in vitro RNA transcription and sequencing
  • Versatile polylinker with flanking EcoR I sites for easy excision of inserts
  • M13 forward and reverse primer sites for sequencing

  • How TA Cloning Works\
  • Taq polymerase has non-template-dependent activity that adds single deoxyadenosine (A) to 3' ends of PCR products
  • Linearized vector supplied in this kit has single 3' deoxythymidine (T) residues
  • Allows PCR inserts to ligate efficiently with vector
  • Kit Configurations
  • TA Cloning Kit is offered in a variety of configurations: with One Shot INVF' Chemically Competent E. coli (K2050-01 and K2050-40), One Shot TOP10F' Chemically Competent E. coli (K2060-01 and K2060-40), and without competent cells (K2750-20 and K2750-40) in 20- and 40-reaction kit sizes

Cloning, PCR Cloning

Spécifications

Spécifications

INVαF ́
TA Cloning
Kit
Linearized pCRII vector, ExpressLink T4 DNA ligase, 5X ExpressLink T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, controls, One Shot INVαF' chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.
One Shot
T7, SP6
pCRII
Chemically Competent E. coli
This kit contains linearized pCR™II vector, ExpressLink™ T4 DNA ligase, 5X ExpressLink™ T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls. Competent cell kits contain One Shot™ chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.

Store One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
PCR Cloning
40 Reactions
Cloning Kit
40 rxns
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.