Learn More
Synthware™ Three Neck Flask, Angled
Three Neck Flask with standard taper outer joints on both center and angled side necks.
Supplier:  Synthware™ F432250
 Afficher plus de versions de ce produit
Numéro de catalogue. 31501254
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • All sizes are hand blown from tubing to ensure uniform wall thickness.
Spécifications
Spécifications
Three Neck Round Bottom Flasks, Angled
Borosilicate Glass (3.3)
Center: 24/40, Side: 19/22
Reusable
Beaded
Round
250 mL
Three-neck Flask
Autoclavable
No
Non-sterile
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS