Learn More
Synthware™ Single Neck Round Bottom Flasks, Full Length Joint
Hand-blown from tubing to ensure uniform wall thickness
Supplier: Synthware™ F302450
Description
SYNTHWARE round bottom boiling flasks with an extended neck and full length standard taper outer joint. All sizes are heavy wall design and are hand blown from borosilicate tubing to ensure uniform wall thickness.Specifications
| Clear | |
| 50 mL | |
| Organic Synthesis |
| Single neck, round bottom flask | |
| Borosilicate Glass (3.3) | |
| 24/40 |