missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Single Neck Round Bottom Flasks, Full Length Joint

Hand-blown from tubing to ensure uniform wall thickness

$52.00 - $289.32

Specifications

Color Clear
Description Single neck, round bottom flask
Material Borosilicate Glass (3.3)
For Use With (Application) Organic Synthesis
View More Specs

Products 16
Catalog Number Mfr. No. Capacity (Metric) Standard Taper Joint Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Standard Taper Joint Price Quantity  
31501103
View Documents
Synthware™
F302425
25 mL 24/40
Each for $52.00
Only null left
 
31501106
View Documents
Synthware™
F302450
50 mL 24/40
Each for $52.00
Only null left
 
31501109
View Documents
Synthware™
F304100
100 mL 24/40
Each for $52.57
Only null left
 
31501110
View Documents
Synthware™
F308100
100 mL 29/42
Each for $64.61
Only null left
 
31501113
View Documents
Synthware™
F304200
200 mL 24/40
Each for $67.46
Only null left
 
31501116
View Documents
Synthware™
F304250
250 mL 24/40
Each for $71.86
Only null left
 
31501117
View Documents
Synthware™
F308250
250 mL 29/42
Each for $75.68
Only null left
 
31501120
View Documents
Synthware™
F304300
300 mL 24/40
Each for $77.75
Only null left
 
31501123
View Documents
Synthware™
F304500
500 mL 24/40
Each for $83.18
Only null left
 
31501124
View Documents
Synthware™
F308500
500 mL 29/42
Each for $85.82
Only null left
 
31501125
View Documents
Synthware™
F303450
500 mL 34/45
Each for $97.93
Only null left
 
31501126
View Documents
Synthware™
F304000
1000 mL 24/40
Each for $100.54
Only null left
 
31501127
View Documents
Synthware™
F30291L
1000 mL 29/42
Each for $109.11
Only null left
 
31501128
View Documents
Synthware™
F30242L
2000 mL 24/40
Each for $266.93
Only null left
 
31501129
View Documents
Synthware™
F30292L
2000 mL 29/42
Each for $272.86
Only null left
 
31501130
View Documents
Synthware™
F30452L
2000 mL 45/50
Each for $289.32
Only null left
 
Description

Description

SYNTHWARE round bottom boiling flasks with an extended neck and full length standard taper outer joint. All sizes are heavy wall design and are hand blown from borosilicate tubing to ensure uniform wall thickness.
Specifications

Specifications

Clear
Single neck, round bottom flask
Borosilicate Glass (3.3)
Organic Synthesis
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'