Learn More
Synthware™ Chromatography Column with PTFE Stopcock
Chromatography Column with PTFE Stopcock for flash chromatography.
$187.17 - $521.13
Specifications
Type Chromatography Column
View More Specs
Products 16
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Inner Size (Metric) Stopcock Diameter (Metric) Outer Price Quantity  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Inner Size (Metric) Stopcock Diameter (Metric) Outer Price Quantity  
31500971
View Documents
Synthware™
C361330
305 mm 10 mm 2 mm bore 13 mm
Each for $231.47
Only null left
31500970
View Documents
Synthware™
C361320
203 mm 10 mm 2 mm bore 13 mm
Each for $195.17
Only null left
31500972
View Documents
Synthware™
C361340
406 mm 10 mm 2 mm bore 13 mm
Each for $256.67
Only null left
31500973
View Documents
Synthware™
C361720
203 mm 13.4 mm 2 mm bore 17 mm
Each for $206.60
Only null left
31500974
View Documents
Synthware™
C361725
254 mm 13.4 mm 2 mm bore 17 mm
Each for $187.17
Only null left
31500975
View Documents
Synthware™
C361730
305 mm 13.4 mm 2 mm bore 17 mm
Each for $242.90
Only null left
31500976
View Documents
Synthware™
C362620
203 mm 20 mm 2 mm bore 26 mm
Each for $206.60
Only null left
31500977
View Documents
Synthware™
C362625
254 mm 20 mm 2 mm bore 26 mm
Each for $224.60
Only null left
31500978
View Documents
Synthware™
C362630
305 mm 20 mm 2 mm bore 26 mm
Each for $242.90
Only null left
31500980
View Documents
Synthware™
C363225
254 mm 26 mm 2 mm bore 32 mm
Each for $290.00
Only null left
31500979
View Documents
Synthware™
C362640
457 mm 20 mm 2 mm bore 26 mm
Each for $304.67
Only null left
31500984
View Documents
Synthware™
C366060
610 mm 53 mm 4 mm bore 60 mm
Each for $521.13
Only null left
31500983
View Documents
Synthware™
C366040
457 mm 53 mm 4 mm bore 60 mm
Each for $441.03
Only null left
31500985
View Documents
Synthware™
C368030
305 mm 73 mm 4 mm bore 80 mm
Each for $462.47
Only null left
31500982
View Documents
Synthware™
C366030
305 mm 53 mm 4 mm bore 60 mm
Each for $364.53
Only null left
31500981
View Documents
Synthware™
C364640
457 mm 40 mm 2 mm bore 46 mm
Each for $384.10
Only null left
Description
Description
  • General purpose chromatography column with PTFE stopcock.
  • With reinforced beaded top.
  • Heavy-wall design increases safety.
Specifications
Specifications
Chromatography Column