Learn More
Synthware™ Chromatography Column with PTFE Stopcock
Chromatography Column with PTFE Stopcock for flash chromatography.
Supplier: Synthware™ C366040
Description
- General purpose chromatography column with PTFE stopcock.
- With reinforced beaded top.
- Heavy-wall design increases safety.
Specifications
| 53 mm | |
| 4 mm bore | |
| 60 mm |
| Chromatography Column | |
| 457 mm |