Learn More
Synthware™ Chromatography Column with PTFE Stopcock
Chromatography Column with PTFE Stopcock for flash chromatography.
Supplier: Synthware™ C364640
Description
- General purpose chromatography column with PTFE stopcock.
- With reinforced beaded top.
- Heavy-wall design increases safety.
Specifications
| 40 mm | |
| 2 mm bore | |
| 46 mm |
| Chromatography Column | |
| 457 mm |