Learn More
Synthware™ Chromatography Column with PTFE Stopcock
Chromatography Column with PTFE Stopcock for flash chromatography.
Supplier:  Synthware™ C361725
 View more versions of this product
Catalog No. 31500974
Only null left
Explore available promotions
Description
Description
  • General purpose chromatography column with PTFE stopcock.
  • With reinforced beaded top.
  • Heavy-wall design increases safety.
Specifications
Specifications
13.4 mm
2 mm bore
17 mm
Chromatography Column
254 mm
Product Suggestions
Product Suggestions