Learn More
Synthware™ Chromatography Column with PTFE Stopcock
Chromatography Column with PTFE Stopcock for flash chromatography.
Supplier:  Synthware™ C361720
 Afficher plus de versions de ce produit
Numéro de catalogue. 31500973
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • General purpose chromatography column with PTFE stopcock.
  • With reinforced beaded top.
  • Heavy-wall design increases safety.
Spécifications
Spécifications
13.4 mm
2 mm bore
17 mm
Chromatography Column
203 mm
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS