Learn More
Student-Grade Tuning Fork Sets
Economical set.
Supplier:  Customer Channels Group (Ccg) 7008STEC
 Afficher plus de versions de ce produit
Numéro de catalogue. S68770
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Eight-piece set includes forks with 256, 288, 320, 341.1, 384, 426.7, 480 and 512Hz
  • 13-Piece set includes forks with 261.6, 277.1, 293.7, 311.1, 329.6, 349.2, 369.9, 392, 415.30, 440, 466.1, 493.9 and 523.2Hz
Spécifications
Spécifications
Tuning Fork Set
Eight piece set includes forks with 256, 288, 320, 341.1, 384, 426.7, 480 and 512Hz
8/Set
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS