Learn More
Spectrum™ Spectra/Por™ 1 RC Dialysis Membrane Tubing 6000 to 8000 Dalton MWCO
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Supplier: Spectrum™ 132670
Description
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.
- Transparent and flexible
- Low molecular weight compounds in aqueous solutions diffuse through membrane
- Higher molecular weights, such as proteins and bacteria, are retained by membrane
- Glycerol preserves membrane and must be removed before use
- Flat sheets and discs available for use with dialysis apparatus
Do not tie knots in tubing; use closures
Spécifications
| Spectra/Por Dialysis Tubing | |
| Regenerated Cellulose | |
| 2.51 in. | |
| 15 m | |
| 4°C to 121°C | |
| Ideal for molecular separation, protein binding studies, electrolyte removal, desalting |
| 6 to 8 kDa | |
| 64 mm | |
| 49.2 ft. | |
| 39.2°F to 249.8°F | |
| Each |