missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por™ 1 RC Dialysis Membrane Tubing 6000 to 8000 Dalton MWCO

Catalog No. p-86909
Change view
Click to view available options
Diameter (Metric):
6.4 mm
14.6 mm
20.4 mm
25.5 mm
32 mm
64 mm
76 mm
Length (Metric):
15 m
15.2 m
30.5 m
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric)
08670C 25.5 mm 30.5 m
08670A 14.6 mm 30.5 m
08670B 20.4 mm 30.5 m
08670F 76 mm 15 m
08670E 64 mm 15 m
086703 6.4 mm 15.2 m
08670D 32 mm 30.5 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 08670C Supplier Spectrum™ Supplier No. 132660
Only null left

Ideal for molecular separation, protein binding studies, electrolyte removal, desalting

Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.

  • Transparent and flexible
  • Low molecular weight compounds in aqueous solutions diffuse through membrane
  • Higher molecular weights, such as proteins and bacteria, are retained by membrane
  • Glycerol preserves membrane and must be removed before use
  • Flat sheets and discs available for use with dialysis apparatus

Do not tie knots in tubing; use closures

Specifications

Product Type Spectra/Por Dialysis Tubing
MWCO 6 to 8 kDa
Material Regenerated Cellulose
Diameter (Metric) 25.5 mm
Diameter (English) 1 in.
Length (English) 100 ft.
Length (Metric) 30.5 m
Temperature Range (English) 39.2°F to 249.8°F
Temperature Range (Metric) 4°C to 121°C
Quantity Each
For Use With (Application) Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Show More Show Less