Learn More
Bel-Art™ Polypropylene Carboy with Spigot
For storing and dispensing large batches of culture media, distilled water, and other solutions.
$630.76 - $760.82
Spécifications
Autoclavable Autoclavable
Certifications/Compliance USP Class VI, 21CFR177.1520, FDA
Closure Material Polypropylene
Closure Type Threaded
Color White
voir les spécifications
Produits 2
Numéro de catalogue Numéro du manufacturier. Capacity (Metric) Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Capacity (Metric) Prix Quantité  
01-189-0110
Afficher les documents Détails de la promotion
Bel-Art™
F118460025
10 L
chaque for $630.76
Il en reste null
01-189-0111
Afficher les documents Détails de la promotion
Bel-Art™
F118460050
20 L
chaque for $760.82
Il en reste null
Description
Description
  • Rugged, FDA-grade polypropylene carboys are 21 CFR 177.1520 compliant for direct contact with foods and beverages
  • USP Class VI certified for demands of biotechnology and pharmaceutical industries
  • Excellent chemical resistance
  • Large handles for ease of transport
  • All components are steam autoclavable at 121°C (250°F): when autoclaving, do not engage cap threads
  • The 83 mm screw cap includes a built-in silicone gasket for positive seal
  • Molded-in bung accepts optional easy-to-use spigot and is located low on carboy for more complete drainage
Spécifications
Spécifications
Autoclavable
Polypropylene
White
For storing and dispensing large batches of culture media, distilled water and other solutions
Polypropylene
Threaded
Cylindrical
USP Class VI, 21CFR177.1520, FDA
Threaded
Reusable
Two large handles, Spigot
Wide
Carboy with Spigot
Non-sterile
Videos
FDS
missing translation for 'documents'
missing translation for 'documents'