missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Tartrate, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$5748.11 - $22108.33
Chemical Identifiers
| Molecular Formula | C4H8Na2O8 |
|---|---|
| Molecular Weight (g/mol) | 230.08 |
| MDL Number | MFCD00150035 |
| InChI Key | FGJLAJMGHXGFDE-UDMGOBQRNA-L |
| PubChem CID | 131855972 |
| SMILES | O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
02004250
|
Avantor
393101 |
500 g | Poly Bottle |
Case of 4 Each for $5,748.11
|
|
||||
|
02004249
|
Avantor J.T.Baker
393105 |
2.5 kg | Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Suitable for Standardization of Karl Fischer Reagents
Chemical Identifiers
| C4H8Na2O8 | |
| MFCD00150035 | |
| 131855972 |
| 230.08 | |
| FGJLAJMGHXGFDE-UDMGOBQRNA-L | |
| O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O |
Specifications
| Crystals | |
| 500 g | |
| MFCD00150035 | |
| 1L = 1.82kg | |
| O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O | |
| 230.08 | |
| 230.08 | |
| Sodium Tartrate, Dihydrate |
| 6106-24-7 | |
| C4H8Na2O8 | |
| Poly Bottle | |
| FGJLAJMGHXGFDE-UDMGOBQRNA-L | |
| (2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate | |
| 131855972 | |
| BAKER ANALYZED™ ACS Reagent |