missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Tartrate, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Description
- Suitable for Standardization of Karl Fischer Reagents
Specifications
Specifications
| CAS | 6106-24-7 |
| Physical Form | Crystals |
| Quantity | 500 g |
| Packaging | Poly Bottle |
| Specific Gravity | 1L = 1.82kg |
| IUPAC Name | (2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate |
| Formula Weight | 230.08 |
| Percent Purity | 100.1% |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Chemical Name or Material | Sodium Tartrate, Dihydrate |