Learn More
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Simplify sampling with easy access
$213.40 - $1,573.63
| Catalog Number | Mfr. No. | For Use With (Equipment) | Type | Holds | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Type | Holds | Price | Quantity & Availability | |||
|
14385361
|
Thermo Scientific™
840298900 |
GENESYS 140/150/180,BioMate160 | Single Cell Sample Holder | 10 mm cuvette (1) |
Each for $459.65
|
|
|||
|
14385362
|
Thermo Scientific™
840299000 |
GENESYS 140/150/180,BioMate160 | Beckman-style Cell Holder | Beckman-style short microcells |
Each for $1,285.17
|
|
|||
|
14385364
|
Thermo Scientific™
840303100 |
GENESYS 140/150/180,BioMate160 | Long Path Cell Holder | 5 to 100mm cylindrical cuvette |
Each for $576.92
|
|
|||
|
14385365
|
Thermo Scientific™
840303200 |
GENESYS 140/150/180,BioMate160 | Sample Holder for Film/Filter | Filter/lenses, 10 mm thick |
Each for $860.69
|
|
|||
|
14385366
|
Thermo Scientific™
840303300 |
GENESYS 140/150/180,BioMate160 | Disposable Microcell Holder | 50 μL microcells, Uvettes |
Each for $1,200.75
|
|
|||
|
14385376
|
Thermo Scientific™
840303800 |
GENESYS 140/150/180,BioMate160 | Long Path Cell Holder | 10 to 100 mm cuvette (1) |
Each for $675.42
|
|
|||
|
14385241
|
Thermo Scientific™
335079000 |
Spectronic200,GENESYS 30/40/50 | Single Cell Sample Holder | Square 10 mm PL Cuvette |
Each for $1,573.63
|
|
|||
|
14385477
|
Thermo Scientific™
335917000 |
GENESYS 30/40/50 | Indiv Adjustable Filter Holder | Filters up to 10 mm thick |
Each for $1,035.00
|
|
|||
|
14380443
|
Thermo Scientific™
840277500 |
GENESYS 30/40/50 | Test Tube and Vial Holder | 12 mm to 25 mm tubes and vials |
Each for $673.08
|
|
|||
|
14380444
|
Thermo Scientific™
840277600 |
GENESYS 30/40/50 | Test Tube Cap | Tall tubes, vials (lid open) |
Each for $443.25
|
|
|||
|
14380445
|
Thermo Scientific™
840277700 |
GENESYS 30/40/50 | Long Path Cell Holder | 20 to 100 mm cuvette (1) |
Each for $609.75
|
|
|||
|
14380446
|
Thermo Scientific™
840277800 |
GENESYS 30/40/50 | Long Path Cell Holder | 5 to 100 mm cuvette (1) |
Each for $551.12
|
|
|||
|
14380447
|
Thermo Scientific™
840278100 |
GENESYS 30/40/50 | Sample Holder for Film/Filter | Filter/lenses, 6 mm thick |
Each for $893.52
|
|
|||
|
14380449
|
Thermo Scientific™
840278200 |
GENESYS 30/40/50 | Hose Entry/Exit Kit | Entry and exit hoses (incl) |
Each for $825.52
|
|
|||
|
14385478
|
Thermo Scientific™
335911000 |
4-position carousel base | Cylindrical Cell Holder | 5 - 100 mm cylindrical cells |
Each for $553.46
|
|
|||
|
14385406
|
Thermo Scientific™
336028000 |
GENESYS 30/40/50 | Single Cell Sample Holder | Single-cell cuvette, 10mm tube |
Each for $213.40
|
|
|||
|
14380448
|
Thermo Scientific™
840278600 |
GENESYS 30/40/50 | Single Cell Sample Holder | 10 mm cuvette (1) |
Each for $433.87
|
|
|||
Description
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.
We provide single-cell solutions for measuring different types of samples.
Get the most from your measurments with accessories for the GENESYS Vis/UV-Vis Spectrophotometer family. Simplify sampling with easy access, flexibility and choices to fit your varied testing needs.