Learn More
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Simplify sampling with easy access
Supplier:  Thermo Scientific™ 840277600
 View more versions of this product
Catalog No. 14380444
Only null left
Description
Description
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.
We provide single-cell solutions for measuring different types of samples.
Get the most from your measurments with accessories for the GENESYS Vis/UV-Vis Spectrophotometer family. Simplify sampling with easy access, flexibility and choices to fit your varied testing needs.
Specifications
Specifications
Test-tube cap covers test-tubes up to 15 cm/6 inches tall to allow measurements while the sample compartment lid is open.
Measurements in 6 in. tall test-tubes, especially in coal mining and agricultural applications.
Tall tubes, vials (lid open)
Each
Test Tube Cap
GENESYS 30/40/50
One cap to cover a tall test tube.
Requires 840-277500 Test-Tube Holder
Product Suggestions
Product Suggestions