Learn More
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Simplify sampling with easy access
Supplier:  Thermo Scientific™ 840278200
 View more versions of this product
Catalog No. 14380449
Only null left
Description
Description
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.
We provide single-cell solutions for measuring different types of samples.
Get the most from your measurments with accessories for the GENESYS Vis/UV-Vis Spectrophotometer family. Simplify sampling with easy access, flexibility and choices to fit your varied testing needs.
Specifications
Specifications
Accessory Kit for Hose Entry/Exit
Thermostatting samples with flowing water. Flow-cell applications with an external pump or sipper.
Entry and exit hoses (incl)
Requires suitable cell holder such as thermostatted cell holder 335079-000 or standard 10 mm square cell holder 336028-000
Hose Entry/Exit Kit
GENESYS 30/40/50
Sample compartment and sample compartment lid, hoses
Product Suggestions
Product Suggestions