Learn More
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Simplify sampling with easy access
Supplier:  Thermo Scientific™ 840278100
 View more versions of this product
Catalog No. 14380447
Only null left
Description
Description
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.
We provide single-cell solutions for measuring different types of samples.
Get the most from your measurments with accessories for the GENESYS Vis/UV-Vis Spectrophotometer family. Simplify sampling with easy access, flexibility and choices to fit your varied testing needs.
Specifications
Specifications
Sample holder for film/filter accommodates filters/lenses up to 50 mm long, 80 mm tall and 6 mm thick. Comes assembled in a removable sample compartment liner.
Measuring transmitting solid samples such as glass or plastic panels, optical filters and specialty films on glass or plastic substrates.
Filter/lenses, 6 mm thick
Sample Holder for Film/Filter
GENESYS 30/40/50
Product Suggestions
Product Suggestions