Learn More
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Simplify sampling with easy access
Supplier:  Thermo Scientific™ 840303800
 View more versions of this product
Catalog No. 14385376
Only null left
Description
Description
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.
We provide single-cell solutions for measuring different types of samples.
Get the most from your measurments with accessories for the GENESYS Vis/UV-Vis Spectrophotometer family. Simplify sampling with easy access, flexibility and choices to fit your varied testing needs.
Specifications
Specifications
Long path rectangular cell holder
Measurements of solutions with low absorbance values
10 to 100 mm cuvette (1)
Long Path Cell Holder
GENESYS 140/150/180,BioMate160
Rectangular
Product Suggestions
Product Suggestions