missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, 0.0141 N (0.0141 M), 1 mL = 0.5 mg Cl-, Ricca Chemical

Supplier:  Ricca Chemical Company R68600001C

 Afficher plus de versions de ce produit

Numéro de catalogue. R68600001C


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Silver Nitrate
98.0,0.23
AgNO3
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
[Ag+].[O-][N+]([O-])=O
169.87
CHEBI:32130
Amber Glass Bottle
∼6
ear colorless liquid
7761-88-8,7732-18-5
100.0,0.24
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
24470
Laboratory
APHA for Chloride
1 L

Identifiants chimiques

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver(1+) nitrate
AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions