missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, 0.0141 N (0.0141 M), 1 mL = 0.5 mg Cl-, Ricca Chemical
$112.34 - $1432.31
Chemical Identifiers
| CAS | 7761-88-8 |
|---|---|
| Molecular Formula | AgNO3 |
| Molecular Weight (g/mol) | 169.87 |
| MDL Number | MFCD00003414 |
| InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
| Synonym | silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
| PubChem CID | 24470 |
| ChEBI | CHEBI:32130 |
| IUPAC Name | silver(1+) nitrate |
| SMILES | [Ag+].[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
6860-16
|
Ricca Chemical Company
686016 |
500 mL | Amber Poly Bottle |
Each for $112.34
|
|
||||
|
R68600001C
|
Ricca Chemical Company
R68600001C |
1 L | Amber Glass Bottle |
Each for $371.31
|
|
||||
|
6860-32
|
Ricca Chemical Company
686032 |
1 L | Amber Poly Bottle |
Each for $120.66
|
|
||||
|
6860-1
|
Ricca Chemical Company
68601 |
4 L | Black Poly Bottle |
Each for $283.75
|
|
||||
|
68602-5
|
Ricca Chemical Company
686025 |
10 L | Cubitainer |
Each for $811.31
|
|
||||
|
6860-5
|
Ricca Chemical Company
68605 |
20 L | Cubitainer |
Each for $1,432.31
|
|
||||
Chemical Identifiers
| 7761-88-8 | |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| 24470 | |
| silver(1+) nitrate |
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| CHEBI:32130 | |
| [Ag+].[O-][N+]([O-])=O |
Specifications
| Colorless | |
| 98.0,0.23 | |
| AgNO3 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| 24470 | |
| Laboratory | |
| APHA for Chloride | |
| 500 mL | |
| 0.0141N,0.0141M |
| 7761-88-8,7732-18-5 | |
| 100.0,0.24 | |
| MFCD00003414 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| 1 mL = 0.5 mg Cl | |
| CHEBI:32130 | |
| Amber Poly Bottle | |
| ∼6 | |
| Liquid | |
| Silver Nitrate |