Learn More
Restek™ Stabilwax™-DB Capillary Columns DFS Item
Application-specific columns that do not require priming; for underivatized amines and other basic compounds including alkylamines, diamines, triamines, nitrogen-containing heterocyclics
Supplier:  Restek™ RK10854
 Afficher plus de versions de ce produit
Numéro de catalogue. 06709220
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Designed specifically to reduce adsorption and improve response of basic compounds such as alkylamines, diamines, triamines, and nitro-containing heterocycles
  • Crossbond™ Carbowax™ technology provides low bleed, increased column life, and solvent rinsability
  • Eliminate need for derivatization
  • No column priming required
  • Tested with a modified Grob test mixture that includes amine compounds
  • Include individual test chromatograph
Spécifications
Spécifications
0.32 mm
Stabilwax™-DB
Triamines Analysis, Alkylamines, Diamines, Triamines, Nitrogen Containing Heterocyclics Analysis
40°C to 210/220°C
30 m
Polar
Amines and other basic compound
220°C
1 μm
1 μm
Stainless Steel
Fused Silica
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS