Learn More
Restek™ Rtx™-CLPesticides2 Capillary Columns DFS Item
Application-specific columns for organochlorine pesticides and herbicides
Supplier:  Restek™ RK11324
 Afficher plus de versions de ce produit
Numéro de catalogue. 06709442
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Specially designed to overcome the coelutions and analyte breakdown typically encountered in chlorinated pesticide analyses for EPA Methods 8081, 608, and CLP
  • Ideal for high sensitivity GC-ECD or GC-MS analyses
  • Baseline separations in less than 10 minutes
  • Low bleed
  • Stable from 320°C to 340°C
  • Analyze EPA Method 8081B, 8082A, 8151A, 504.1, 515, 508.1, and 552.2 compounds without time-consuming column changes
Spécifications
Spécifications
0.32 mm
350°C
-60°C to +320°C
30 m
Fused Silica
Rtx™-CLPII
Organochlorine Pesticides Analysis, Organochlorine Pesticides, Herbicides Analysis
0.25 μm
Stainless Steel
Ea.
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS