missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ Class A Graduated Cylinders with Double Metric Scale, TD
GREENER_CHOICE

Calibrated To Deliver

Supplier:  Corning™ 3026250

 Afficher plus de versions de ce produit

Numéro de catalogue. 08-549-11K


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Calibrated To Deliver

Cylinders are calibrated 'To Deliver' in accordance with ASTM E-1272 and ASTM E-542. The double scale, numbered up and down, is in durable white enamel. Supplied with shock absorbing poly­ethylene bumper guards.

Compliance

In accordance with ASTM™ E 1272 and ASTM E 542.

Greener Choice Claims
  • Environmental benefits include:
    • Énergie renouvelable
Spécifications

Spécifications

Round
Graduated Cylinder
To Deliver
Blue
Hexagonal
340 mm
1.5 in.
250 mL
Dual
Autoclavable
±1 mL
13.3 in.
Non-sterile
ASTM E1272, ASTM E542, ASTM E694
A
Borosilicate Glass
40 mm
Bumper Guard
2 mL
2 mL
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Accessoires
Promotions

Promotions