missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotix Pure uTIP™ Bulk Non-sterile Pipette Tips

Pure uTIP™ Low Retention Non-Filter Universal Pipette Tips, Standard, Bulk are packaged in re-sealable bags making storage and racking simple. RNase, DNase, Pyrogen, and PCR Inhibitor free.
Supplier: Biotix 63305423
Description
- Biotix™ uTIP™ Low Retention Non-Filter Universal Pipette Tips, Standard, Bulk
- Packaged in re-sealable bags making storage and racking simple.
- Biotix™ uTIP™ Pure resin Non-Filter Universal Pipette Tips, Bulk are RNase, DNase, Pyrogens, and PCR Inhibitors free.
- Fits leading pipette brands and improves ergonomics by lowering insertion and ejection forces
- Achieved through patented technologies including FlexFit and Blade
- Pure resin made of virgin polypropylene
- Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
- Re-sealable bag
- Tips must be racked by hand
Greener Choice Claims
- Environmental benefits include:
- Recyclable
- Certifications et labels de qualité
Spécifications
300 μL | |
Non-sterile | |
Autoclavable | |
Hinged Rack | |
960 Pk. | |
Green | |
Universal Fit, Universal Fit Multichannel | |
uTip | |
PCR Ready |
Non-filtered | |
Clear | |
Universal | |
Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point | |
Pipette Tips | |
Single-use | |
Yes | |
Polypropylene |
Promotions
-
Biotix
Obtenez 1 caisse de pointes de pipette avec votre achat
Achetez trois caisses admissibles de pointes de pipette BioTix et recevez une quatrième caisse de pointes.Expire: 08/31/25 Redemption Only
This product contains no safety hazards