missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Tissue Strainers, 250 μm, 2.5 mL

Rapidly separate cells or cell lysate from tissue debris after mechanical disruption using a 250-micron mesh filter and gravity or gentle centrifugation.

Supplier:  Thermo Scientific™ 87791

Numéro de catalogue. PI87791

Il en reste null
Découvrez plus d'offres spéciales

Description

Description

The Thermo Scientific™ Pierce Tissue Strainer fits standard 15mL conical tubes for quick separation of cells or cell lysate from tissue debris through a 250μm mesh filter after mechanical disruption using gravity or gentle centrifugation.

The easy-to-use Pierce Tissue Strainers are made of strong nylon and enable rapid isolation cells or uniform protein extracts from up to 2.5mL of tissue sample per load. The tissue strainers fit into standard 15mL conical tubes, making the processing and recovery of samples simple and reliable.

Highlights:

  • Convenient remove tissue debris from tissue homogenate within minutes using gravity filtration or gentle centrifugation

  • Compatible unique design fits into standard 15mL conical tubes

  • Versitle remove debris from a variety of tissues types, including liver, heart, brain, kidney, lung and spleen

Recommended for:

Use with the Subcellular Protein Fractionation Kit for Tissues and other protein extraction reagent protocols that start with tissue samples; Improve uniformity of tissue homogenates to facilitate sample handling; Protect downstream instrumentation and devices by removing tissue clumps and debris that may foul injection ports and columns

Spécifications

Spécifications

Tissue Strainer
250μM Nylon Mesh on Inert Plastic Frame
Pierce
50 Units
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.