missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus ICP Standard, 10,000 ppm P in H2O, Ricca Chemical

Supplier:  Ricca Chemical Company PP10KW500

 Afficher plus de versions de ce produit

Numéro de catalogue. PP10KW-500

Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Phosphorus ICP Standard
7722-76-1,7732-18-5
10,000ppm P
1 mL = 10 mg P
H6NO4P
Colorless
Standard
N.OP(O)(O)=O
115.03
24402
Natural Poly Bottle
94.0,3.51
98.0,3.65
Liquid
MFCD00003396
Approximately 0°C
LFVGISIMTYGQHF-UHFFFAOYSA-N
phosphoric acid amine
500 mL
CHEBI:62982

Identifiants chimiques

7722-76-1
115.03
LFVGISIMTYGQHF-UHFFFAOYSA-N
CHEBI:62982
N.OP(O)(O)=O
H6NO4P
MFCD00003396
24402
phosphoric acid amine
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions