missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus ICP Standard, 10,000 ppm P in H2O, Ricca Chemical

Catalog No. p-3252074
Change view
Click to view available options
Quantity:
50 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PP10KW50 50 mL Natural Poly Bottle
PP10KW500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PP10KW50 Supplier Ricca Chemical Company Supplier No. PP10KW50
Only null left

Chemical Identifiers

CAS 7722-76-1 , 7732-18-5
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name phosphoric acid amine
SMILES N.OP(O)(O)=O

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 10,000ppm P
CAS Min % 94.0, 3.51
CAS Max % 98.0, 3.65
Equivalence 1 mL = 10 mg P
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Type Standard
Quantity 50 mL
Chemical Name or Material Phosphorus ICP Standard
Show More Show Less