Learn More
p-Tolyl isocyanate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 162520050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
p-Tolyl isocyanate | |
1.0560g/mL | |
66°C | |
99% | |
C8H7NO | |
CH3C6H4NCO | |
12,955 | |
p-tolyl isocyanate, 4-tolyl isocyanate, 4-methylphenyl isocyanate, p-isocyanatotoluene, 4-isocyanatotoluene, p-toluene isocyanate, p-methylphenyl isocyanate, benzene, 1-isocyanato-4-methyl, p-tolylisocyanate, 1-isocyanato-4-methyl-benzene | |
CC1=CC=C(C=C1)N=C=O | |
133.15 | |
CHEBI:53732 | |
99% |
622-58-2 | |
70.0°C to 72.0°C (10.0mmHg) | |
Authentic | |
Glass bottle | |
1.5301 to 1.5321 | |
5g | |
1.056 | |
MGYGFNQQGAQEON-UHFFFAOYSA-N | |
1-isocyanato-4-methylbenzene | |
69325 | |
133.15 |
Identifiants chimiques
622-58-2 | |
133.15 | |
p-tolyl isocyanate, 4-tolyl isocyanate, 4-methylphenyl isocyanate, p-isocyanatotoluene, 4-isocyanatotoluene, p-toluene isocyanate, p-methylphenyl isocyanate, benzene, 1-isocyanato-4-methyl, p-tolylisocyanate, 1-isocyanato-4-methyl-benzeneAfficher plus de résultats | |
CHEBI:53732 | |
CC1=CC=C(C=C1)N=C=O |
C8H7NO | |
MGYGFNQQGAQEON-UHFFFAOYSA-N | |
69325 | |
1-isocyanato-4-methylbenzene |
Sécurité et manipulation
GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Very toxic
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 210-743-4
RUO â Research Use Only