Learn More
Invitrogen™ Novex™ Tricine SDS Sample Buffer (2X)
Use with Novex Tricine Gels
Supplier: Invitrogen™ LC1676
Description
In the tricine system developed by Schaegger and von Jagow (1), tricine replaces glycine in the running buffer. This results in more efficient stacking and destacking of low molecular weight proteins and higher resolution of smaller peptides.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice
Spécifications
| Sample Loading Buffer | |
| Liquid | |
| 20 mL |
| 2X | |
| Novex |
For Research Use Only. Not for use in diagnostic procedures.