missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[NH2Me2][(RuCl((R)-binap))2(mu-Cl)3], TCI America™
Supplier: TCI America N10151G
Spécifications
| [NH2Me2][(RuCl((R)-binap))2(mu-Cl)3] | |
| C90H75Cl5NP4Ru2+ | |
| 1 g | |
| NVDGOPZSESFSJS-UHFFFAOYSA-M | |
| [1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;N-methylmethanamine;ruthenium;ruthenium(2+);chloride;tetrahydrochloride | |
| 131675866 | |
| Crystalline Powder |
| 199684-47-4 | |
| MFCD09753033 | |
| Dimethylammonium Dichlorotri(mu-chloro)bis[(R)-(+)-2,2′C-bis(diphenylphosphino)-1,1′C-binaphthyl]diruthenate(II) | |
| CNC.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.Cl.Cl.Cl.Cl.[Cl-].[Ru].[Ru+2] | |
| 1673.882 | |
| 1670.86 |
Identifiants chimiques
| 199684-47-4 | |
| 1673.882 | |
| NVDGOPZSESFSJS-UHFFFAOYSA-M | |
| 131675866 | |
| CNC.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.Cl.Cl.Cl.Cl.[Cl-].[Ru].[Ru+2] |
| C90H75Cl5NP4Ru2+ | |
| MFCD09753033 | |
| Dimethylammonium Dichlorotri(mu-chloro)bis[(R)-(+)-2,2′C-bis(diphenylphosphino)-1,1′C-binaphthyl]diruthenate(II) | |
| [1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;N-methylmethanamine;ruthenium;ruthenium(2+);chloride;tetrahydrochloride |
Sécurité et manipulation
missing translation for 'tsca' : No