missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[NH2Me2][(RuCl((R)-binap))2(mu-Cl)3], TCI America™
$80.16 - $260.97
Chemical Identifiers
| CAS | 199684-47-4 |
|---|---|
| Molecular Formula | C90H75Cl5NP4Ru2+ |
| Molecular Weight (g/mol) | 1673.882 |
| MDL Number | MFCD09753033 |
| InChI Key | NVDGOPZSESFSJS-UHFFFAOYSA-M |
| Synonym | Dimethylammonium Dichlorotri(mu-chloro)bis[(R)-(+)-2,2′C-bis(diphenylphosphino)-1,1′C-binaphthyl]diruthenate(II) |
| PubChem CID | 131675866 |
| IUPAC Name | [1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;N-methylmethanamine;ruthenium;ruthenium(2+);chloride;tetrahydrochloride |
| SMILES | CNC.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.Cl.Cl.Cl.Cl.[Cl-].[Ru].[Ru+2] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
N1015200MG
|
TCI America
N1015200MG |
200 mg |
Each for $80.16
|
|
|||||
|
N10151G
|
TCI America
N10151G |
1 g |
Each for $260.97
|
|
|||||
Chemical Identifiers
| 199684-47-4 | |
| 1673.882 | |
| NVDGOPZSESFSJS-UHFFFAOYSA-M | |
| 131675866 | |
| CNC.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.Cl.Cl.Cl.Cl.[Cl-].[Ru].[Ru+2] |
| C90H75Cl5NP4Ru2+ | |
| MFCD09753033 | |
| Dimethylammonium Dichlorotri(mu-chloro)bis[(R)-(+)-2,2′C-bis(diphenylphosphino)-1,1′C-binaphthyl]diruthenate(II) | |
| [1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;N-methylmethanamine;ruthenium;ruthenium(2+);chloride;tetrahydrochloride |
Specifications
| 199684-47-4 | |
| MFCD09753033 | |
| Dimethylammonium Dichlorotri(mu-chloro)bis[(R)-(+)-2,2′C-bis(diphenylphosphino)-1,1′C-binaphthyl]diruthenate(II) | |
| CNC.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.Cl.Cl.Cl.Cl.[Cl-].[Ru].[Ru+2] | |
| 1673.882 | |
| 1670.86 | |
| [NH2Me2][(RuCl((R)-binap))2(mu-Cl)3] |
| C90H75Cl5NP4Ru2+ | |
| 200 mg | |
| NVDGOPZSESFSJS-UHFFFAOYSA-M | |
| [1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;N-methylmethanamine;ruthenium;ruthenium(2+);chloride;tetrahydrochloride | |
| 131675866 | |
| Crystalline Powder |
Safety and Handling
TSCA : No